Learn More
Agilent Technologies ADEASY ADENOVIRAL VECTOR SYSA
Catalog No. 50125391
Encompass
AdEasy Adenoviral Vector System employes Homologous recombination in E.coli. It has High-level protein expression. Infects both dividing and non-dividing cells with 100% transduction efficiency. Vitality hrGFP reporter is available.
Catalog No. 50-125-391 Supplier Agilent Technologies Supplier No. 240009
AdEasy Adenoviral Vector System employes Homologous recombination in E.coli. It has High-level protein expression. Infects both dividing and non-dividing cells with 100% transduction efficiency. Vitality hrGFP reporter is available.