Learn More
Agilent Technologies ADEASY XL ADENOVIRAL VECT SYS
Catalog No. 50125392
Encompass
AdEasy XL Adenoviral Vector System. Contains:20microgram pShuttle-CMV vector, 10microgram pShuttle-CMV-lacZ ctrl vector,BJ-5183-Ad1 electrocompetent cells, 10microliter Transformation ctrl plasmid, XL10-Gold ultracompetent cells,10microlite
Catalog No. 50-125-392 Supplier Agilent Technologies Supplier No. 240010
AdEasy XL Adenoviral Vector System. Contains:20microgram pShuttle-CMV vector, 10microgram pShuttle-CMV-lacZ ctrl vector,BJ-5183-Ad1 electrocompetent cells, 10microliter Transformation ctrl plasmid, XL10-Gold ultracompetent cells,10microlite