Learn More
Cayman Chemical Adefovir dipivoxil, Purity: ≥98%, CAS: 142340-99-6, 100 mg
SDP
Catalog No. 502938557
Encompass_Preferred
A prodrug form of adefovir; inhibits HBV DNA synthesis in HepG2 2.2.15 cells infected with HBV (EC50 = 0.517 μM); cytotoxic to HepG2 2.2.15 cells (CC50 = 540 μM); inhibits HBV replication in the liver of HBV transgenic mice (ED50 = 0.2 μmol/kg per day)
Catalog No. 50-293-8557 Supplier Cayman Chemical Supplier No. 28373100 MG
A prodrug form of adefovir; inhibits HBV DNA synthesis in HepG2 2.2.15 cells infected with HBV (EC50 = 0.517 μM); cytotoxic to HepG2 2.2.15 cells (CC50 = 540 μM); inhibits HBV replication in the liver of HBV transgenic mice (ED50 = 0.2 μmol/kg per day)