Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adefovir Dipivoxil 98.0+%, TCI America™
SDP

Catalog No. A24141G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24141G 1 g
A2414100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24141G Supplier TCI America Supplier No. A24141G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 142340-99-6
Molecular Formula C20H32N5O8P
Molecular Weight (g/mol) 501.48
MDL Number MFCD00869897
InChI Key WOZSCQDILHKSGG-UHFFFAOYSA-N
Synonym 9-[2-[Bis(pivaloyloxymethoxy)phosphorylmethoxy]ethyl]adenine
PubChem CID 60871
IUPAC Name [({[2-(6-amino-9H-purin-9-yl)ethoxy]methyl}({[(2,2-dimethylpropanoyl)oxy]methoxy})phosphoryl)oxy]methyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OCOP(=O)(COCCN1C=NC2=C(N)N=CN=C12)OCOC(=O)C(C)(C)C

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 1 g
Formula Weight 501.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Adefovir Dipivoxil
RTECSNumber UA2459362
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.