Learn More
Medchemexpress LLC Adelmidrol | 1675-66-7 | 98.0% | 250 MG
SDP
Catalog No. 5000414481
Encompass_Preferred
Adelmidrol exerts important anti-inflammatory effects that are partly dependent on PPARγ. It reduces NF-κB translocation and COX-2 expression.
  • Palmitoylethanolamide analogue.
  • Reduces NF-κB translocation, COX-2, and p-ERK expression.
  • Reduces proinflammatory cytokine release.
  • Reduces the incidence of nitrotyrosine and poly(ADP)ribose in the colon.
  • Reduces the degree and severity of macroscopic and histologic signs of colon injury in vivo.
  • Significantly reduces body weight loss in an inflammatory bowel disease model.
  • Significantly reduces polymorphonuclear cell infiltration in inflamed colon.
  • Potential pharmacological treatment for inflammatory bowel disease.
Catalog No. 50-004-14481 Supplier Medchemexpress LLC Supplier No. 250MGHYB1026
Adelmidrol exerts important anti-inflammatory effects that are partly dependent on PPARγ. It reduces NF-κB translocation and COX-2 expression.
  • Palmitoylethanolamide analogue.
  • Reduces NF-κB translocation, COX-2, and p-ERK expression.
  • Reduces proinflammatory cytokine release.
  • Reduces the incidence of nitrotyrosine and poly(ADP)ribose in the colon.
  • Reduces the degree and severity of macroscopic and histologic signs of colon injury in vivo.
  • Significantly reduces body weight loss in an inflammatory bowel disease model.
  • Significantly reduces polymorphonuclear cell infiltration in inflamed colon.
  • Potential pharmacological treatment for inflammatory bowel disease.