Learn More
Selleck Chemical LLC Ademetionine 25mg 29908-03-0 AdoMet, S-Adenosylmethionine, SAMe
SDP
Supplier:  Selleck Chemical LLC S510925MG
Encompass_Preferred
Ademetionine (AdoMet, S-Adenosylmethionine, SAMe), also known as SAMe, is a specific form of the amino acid methionine known as S-adenosyl-methionine. It is essential for the formation of glutathione, a water-soluble peptide that helps the body fight free radicals. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8405