Learn More
Cayman Chemical AdenIn15N5hydroCLOrID h 500ug
SDP
Catalog No. 502943706
Encompass_Preferred
An internal standard for the quantification of adenine by GC- or LC-MS
Catalog No. 50-294-3706 Supplier Cayman Chemical Supplier No. 41181500 UG
An internal standard for the quantification of adenine by GC- or LC-MS