Learn More
Research Products International Corp Adenine [6-Aminopurine] [Vitamin B4], 100 Grams
SDP
Supplier:  Research Products International Corp A11500100.0
Encompass_Preferred
A nucleobase with a variety of roles in biochemistry including cellular respiration, in the form of both the energy-rich adenosine triphosphate and the cofactors nicotinamide adenine dicucleotide and flavin adenine dinucleotide, and protein synthesis, as a chemical component of DNA and RNA.
Catalog No. 50-488-444