Learn More
Medchemexpress LLC Adenine | 73-24-5 | MFCD00005701 | ≥99% | 5 G
SDP
Catalog No. 501639211
Encompass_Preferred
Adenine (6-Aminopurine) is a purine and one of the four nucleobases found in the nucleic acid of DNA. It serves as a chemical component of both DNA and RNA. Adenine plays a vital role in biochemistry, involved in cellular respiration, the formation of both ATP and cofactors (NAD and FAD), and protein synthesis.
  • One of the four nucleobases found in DNA
  • Serves as a chemical component of DNA and RNA
  • Plays a vital role in cellular respiration
  • Involved in the formation of ATP and cofactors (NAD and FAD)
  • Participates in protein synthesis
Catalog No. 50-163-9211 Supplier Medchemexpress LLC Supplier No. HYB01525G
Adenine (6-Aminopurine) is a purine and one of the four nucleobases found in the nucleic acid of DNA. It serves as a chemical component of both DNA and RNA. Adenine plays a vital role in biochemistry, involved in cellular respiration, the formation of both ATP and cofactors (NAD and FAD), and protein synthesis.
  • One of the four nucleobases found in DNA
  • Serves as a chemical component of DNA and RNA
  • Plays a vital role in cellular respiration
  • Involved in the formation of ATP and cofactors (NAD and FAD)
  • Participates in protein synthesis