Learn More
Medchemexpress LLC Adenine-13C | 86967-48-8 | 99.1% | 1 MG
SDP
Catalog No. 5000406266
Encompass_Preferred
Adenine-13C is the 13C labeled Adenine. As 6-Aminopurine, it is a purine and one of the four nucleobases found in the nucleic acid of DNA and RNA. This compound plays a significant role in biochemistry, particularly in cellular respiration, forming both ATP and cofactors (NAD and FAD), and protein synthesis.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06266 Supplier Medchemexpress LLC Supplier No. HYB0152S11MG
Adenine-13C is the 13C labeled Adenine. As 6-Aminopurine, it is a purine and one of the four nucleobases found in the nucleic acid of DNA and RNA. This compound plays a significant role in biochemistry, particularly in cellular respiration, forming both ATP and cofactors (NAD and FAD), and protein synthesis.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS