Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenine-13C | 86967-48-8 | 99.1% | 136.12 g/mol | C5H5N5 (13C-labeled) | 10 MG
SDP

Catalog No. 5000298249
Encompass_Preferred

Adenine-13C is a stable isotope-labeled adenine (single 13C) supplied as a white to off-white solid for use in mass spectrometry, metabolic tracing, and biochemical assays as an internal standard or tracer. It is intended for research use and should be stored protected from light under inert atmosphere.

  • Stable isotope-labeled adenine with a single 13C atom.
  • High purity suitable for analytical applications and tracer studies.
  • White to off-white solid, easy to weigh and handle.
  • Recommended storage: protected from light, under nitrogen, refrigerated or frozen for long-term stability.
  • Applications: mass spectrometry internal standard, metabolic flux analysis, and biochemical research.

Catalog No. 50-002-98249 Supplier Medchemexpress LLC Supplier No. HYB0152S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenine-13C is a stable isotope-labeled adenine (single 13C) supplied as a white to off-white solid for use in mass spectrometry, metabolic tracing, and biochemical assays as an internal standard or tracer. It is intended for research use and should be stored protected from light under inert atmosphere.

  • Stable isotope-labeled adenine with a single 13C atom.
  • High purity suitable for analytical applications and tracer studies.
  • White to off-white solid, easy to weigh and handle.
  • Recommended storage: protected from light, under nitrogen, refrigerated or frozen for long-term stability.
  • Applications: mass spectrometry internal standard, metabolic flux analysis, and biochemical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.