Learn More
Medchemexpress LLC Adenine-13c | 86967-48-8 | 99.1% | 25 MG
SDP
Catalog No. 5000407438
Encompass_Preferred
Adenine-13C is a 13C labeled Adenine, a purine and one of the four nucleobases found in DNA and RNA. It plays a vital role in biochemistry, specifically in cellular respiration, the formation of ATP and cofactors like NAD and FAD, and protein synthesis. It can be utilized as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • 13C labeled adenine
  • Purity 99.06%
  • Suitable for use as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS techniques
Catalog No. 50-004-07438 Supplier Medchemexpress LLC Supplier No. 25MGHYB0152S1
Adenine-13C is a 13C labeled Adenine, a purine and one of the four nucleobases found in DNA and RNA. It plays a vital role in biochemistry, specifically in cellular respiration, the formation of ATP and cofactors like NAD and FAD, and protein synthesis. It can be utilized as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • 13C labeled adenine
  • Purity 99.06%
  • Suitable for use as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, or LC-MS techniques