Learn More
Medchemexpress LLC Adenine-13C | 86967-48-8 | 99.1% | 5 MG
SDP
Catalog No. 5000406228
Encompass_Preferred
Adenine-13C is the 13C labeled Adenine. Adenine (6-Aminopurine), a purine, is one of the four nucleobases in the nucleic acid of DNA. It acts as a chemical component of DNA and RNA and plays an important role in biochemistry involved in cellular respiration, the form of both ATP and the cofactors (NAD and FAD), and protein synthesis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06228 Supplier Medchemexpress LLC Supplier No. HYB0152S15MG
Adenine-13C is the 13C labeled Adenine. Adenine (6-Aminopurine), a purine, is one of the four nucleobases in the nucleic acid of DNA. It acts as a chemical component of DNA and RNA and plays an important role in biochemistry involved in cellular respiration, the form of both ATP and the cofactors (NAD and FAD), and protein synthesis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.