Learn More
Medchemexpress LLC Adenine-15N5 | 73-24-5 | 95.0% | 5 MG
SDP
Catalog No. 5000406252
Encompass_Preferred
This product is a 15N labeled nucleobase, which is one of the four essential components of DNA and RNA. It plays a crucial role in biochemistry, including cellular respiration, the formation of ATP and cofactors (NAD and FAD), and protein synthesis.It is intended for research use only.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can impact pharmacokinetic and metabolic profiles.
Catalog No. 50-004-06252 Supplier Medchemexpress LLC Supplier No. HYB0152S45MG
This product is a 15N labeled nucleobase, which is one of the four essential components of DNA and RNA. It plays a crucial role in biochemistry, including cellular respiration, the formation of ATP and cofactors (NAD and FAD), and protein synthesis.It is intended for research use only.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can impact pharmacokinetic and metabolic profiles.