Learn More
Research Products International Corp Adenine [6-Aminopurine] [Vitamin B4] USP, 25 Grams
SDP
Catalog No. 501368011
Encompass_Preferred
A nucleobase with a variety of roles in biochemistry including cellular respiration, in the form of both the energy-rich adenosine triphosphate and the cofactors nicotinamide adenine dicucleotide and flavin adenine dinucleotide, and protein synthesis, as a chemical component of DNA and RNA.
Catalog No. 50-136-8011 Supplier Research Products International Corp Supplier No. A1150025.0
A nucleobase with a variety of roles in biochemistry including cellular respiration, in the form of both the energy-rich adenosine triphosphate and the cofactors nicotinamide adenine dicucleotide and flavin adenine dinucleotide, and protein synthesis, as a chemical component of DNA and RNA.