Learn More
Medchemexpress LLC Adenine (6-aminopurine) | 73-24-5 | MFCD00041790 | 99.9% | 135.13 g/mol | C5H5N5 | 10 MG
SDP
Catalog No. 5000299046
Encompass_Preferred
Adenine (6-aminopurine) analytical standard supplied as a high-purity reference material for research and analytical applications. It appears as a white to off-white crystalline powder and is provided with supporting documentation for analytical use.
  • High purity: 99.91%.
  • Chemical formula C5H5N5 and molecular weight 135.13 g/mol.
  • Suitable as a reference standard for HPLC, LC-MS, and other analytical assays.
  • Provided with a certificate of analysis and safety data sheet for traceability.
  • Intended for research and analytical use only.
  • Available in small quantities for method development and calibration.
Catalog No. 50-002-99046 Supplier Medchemexpress LLC Supplier No. HYB0152R10MG
Adenine (6-aminopurine) analytical standard supplied as a high-purity reference material for research and analytical applications. It appears as a white to off-white crystalline powder and is provided with supporting documentation for analytical use.
  • High purity: 99.91%.
  • Chemical formula C5H5N5 and molecular weight 135.13 g/mol.
  • Suitable as a reference standard for HPLC, LC-MS, and other analytical assays.
  • Provided with a certificate of analysis and safety data sheet for traceability.
  • Intended for research and analytical use only.
  • Available in small quantities for method development and calibration.