Learn More
Alkali Scientific Adenine [6 Aminopurine] [Vitamin B4], USP, 100 G
SDP
Catalog No. 5000661477
Encompass_Preferred
Adenine (USP Grade), 100 g, is a purine nucleobase essential for plant and microbial tissue culture. As a key building block of nucleic acids, adenine plays a critical role in cellular metabolism and growth. In plant tissue culture, it is often used to enhance the growth of callus tissue and promote the regeneration of shoots and roots. Adenine supports the synthesis of adenine-based nucleotides, which are crucial for cellular functions such as DNA replication and energy transfer. This 100g pack provides an ample supply of high-quality adenine for large-scale experiments and ongoing research projects in plant biotechnology, agriculture, and cell culture studies.
Catalog No. 50-006-61477 Supplier Alkali Scientific Supplier No. NEA001CP
Adenine (USP Grade), 100 g, is a purine nucleobase essential for plant and microbial tissue culture. As a key building block of nucleic acids, adenine plays a critical role in cellular metabolism and growth. In plant tissue culture, it is often used to enhance the growth of callus tissue and promote the regeneration of shoots and roots. Adenine supports the synthesis of adenine-based nucleotides, which are crucial for cellular functions such as DNA replication and energy transfer. This 100g pack provides an ample supply of high-quality adenine for large-scale experiments and ongoing research projects in plant biotechnology, agriculture, and cell culture studies.