Learn More
Alkali Scientific Adenine [6 Aminopurine] [Vitamin B4] USP, 5 Grams
SDP
Catalog No. 5000661465
Encompass_Preferred
Adenine (Vitamin B4) is a purine base crucial for nucleic acid metabolism, widely used in biochemical and molecular biology research. This 5g reagent is essential for studies involving DNA, RNA, and cellular replication. Adenine serves as a key building block in nucleotides and is involved in the synthesis of adenine-containing coenzymes like ATP and NAD. In laboratory settings, Adenine is often used in cell culture media and in experiments exploring gene expression, mutagenesis, and metabolic pathways. It is a critical component in the study of nucleotide biosynthesis and is used in various research applications related to genetics and biochemistry.
Catalog No. 50-006-61465 Supplier Alkali Scientific Supplier No. NEA005CP
Adenine (Vitamin B4) is a purine base crucial for nucleic acid metabolism, widely used in biochemical and molecular biology research. This 5g reagent is essential for studies involving DNA, RNA, and cellular replication. Adenine serves as a key building block in nucleotides and is involved in the synthesis of adenine-containing coenzymes like ATP and NAD. In laboratory settings, Adenine is often used in cell culture media and in experiments exploring gene expression, mutagenesis, and metabolic pathways. It is a critical component in the study of nucleotide biosynthesis and is used in various research applications related to genetics and biochemistry.