Learn More
Alkali Scientific Adenine [6 Aminopurine] [Vitamin B4], USP, 500 G
SDP
Catalog No. 5000661418
Encompass_Preferred
Adenine [6 Aminopurine] [Vitamin B4] is a USP grade reagent available in a 500g bottle. As a purine nucleobase, adenine plays an essential role in nucleic acid metabolism and is commonly used in cell culture media for research in molecular biology and biochemistry. Adenine is a precursor for the synthesis of DNA and RNA and is important for the growth and maintenance of various cell types in vitro. It is widely used in experiments involving cellular processes, including the study of cell growth, gene expression, and enzyme activity. This high-quality reagent is ideal for laboratory applications that require the precise formulation of culture media or for the synthesis of compounds that involve adenine.
Catalog No. 50-006-61418 Supplier Alkali Scientific Supplier No. NEA002CP
Adenine [6 Aminopurine] [Vitamin B4] is a USP grade reagent available in a 500g bottle. As a purine nucleobase, adenine plays an essential role in nucleic acid metabolism and is commonly used in cell culture media for research in molecular biology and biochemistry. Adenine is a precursor for the synthesis of DNA and RNA and is important for the growth and maintenance of various cell types in vitro. It is widely used in experiments involving cellular processes, including the study of cell growth, gene expression, and enzyme activity. This high-quality reagent is ideal for laboratory applications that require the precise formulation of culture media or for the synthesis of compounds that involve adenine.