Learn More
Bioworld Adenine, 25 g
Supplier:  Bioworld 401500001
Encompass_Preferred
Adenine (6-Aminopurine)Synonyms: 6-Aminopurine or Vitamin B4A purine nucleobase and a component of DNAAuxin growth regulatorUltra Pure GradePlant cell culture testedAppearance: SolidBoiling Point : 553.49 °C at 760 mmHg (Predicted)Density : 1.61 g/cm3 (Predicted)Melting Point: >360 °C (lit.)RTECS Number: AU6125000
Catalog No. 50-255-354