Learn More
Apexbio Technology LLC Adenine 73-24-5 1g
SDP
Catalog No. 5000090458
Encompass_Preferred
Adenine (73-24-5) is a selective agonist targeting high-affinity adenine receptors It is designed to activate these receptors thereby mediating cellular signaling cascades and regulating diverse physiological and biochemical pathways Adenine exerts its biological activity primarily through receptor-ligand interactions that trigger downstream intracellular signaling Experimental evidence indicates that adenine exhibits receptor activation with IC50 values typically ranging from 50 to 200 nM dependent upon the specific receptor subtype and assay context Based on these pharmacological properties adenine holds research potential in receptor binding assays pharmacological experiments to elucidate receptor function assess signaling mechanisms and investigate therapeutic interventions targeting adenine-responsive pathways
Catalog No. 50-000-90458 Supplier Apexbio Technology LLC Supplier No. B14681000
Adenine (73-24-5) is a selective agonist targeting high-affinity adenine receptors It is designed to activate these receptors thereby mediating cellular signaling cascades and regulating diverse physiological and biochemical pathways Adenine exerts its biological activity primarily through receptor-ligand interactions that trigger downstream intracellular signaling Experimental evidence indicates that adenine exhibits receptor activation with IC50 values typically ranging from 50 to 200 nM dependent upon the specific receptor subtype and assay context Based on these pharmacological properties adenine holds research potential in receptor binding assays pharmacological experiments to elucidate receptor function assess signaling mechanisms and investigate therapeutic interventions targeting adenine-responsive pathways