Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenine | 73-24-5 | 99.99% | 1 G
SDP

Catalog No. 5000406264
Encompass_Preferred

Adenine (6-Aminopurine) is a purine and one of the four nucleobases found in DNA. It functions as a chemical component of both DNA and RNA, playing a vital role in biochemistry, particularly in cellular respiration where it forms part of ATP and cofactors like NAD and FAD, and in protein synthesis.

  • Chemical component of DNA and RNA
  • Important in cellular respiration, forming ATP, NAD, and FAD
  • Involved in protein synthesis
  • Purity of 99.99%
  • Molecular weight of 135.13
  • Formula C5H5N5
  • White to off-white solid appearance
  • Soluble in DMSO (≥ 10 mg/mL) and H2O (2 mg/mL with warming)
  • Utilized to induce chronic kidney injury models
  • Used in inducing Chronic Kidney Disease (CKD) models in mice and rats

Catalog No. 50-004-06264 Supplier Medchemexpress LLC Supplier No. HYB01521G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenine (6-Aminopurine) is a purine and one of the four nucleobases found in DNA. It functions as a chemical component of both DNA and RNA, playing a vital role in biochemistry, particularly in cellular respiration where it forms part of ATP and cofactors like NAD and FAD, and in protein synthesis.

  • Chemical component of DNA and RNA
  • Important in cellular respiration, forming ATP, NAD, and FAD
  • Involved in protein synthesis
  • Purity of 99.99%
  • Molecular weight of 135.13
  • Formula C5H5N5
  • White to off-white solid appearance
  • Soluble in DMSO (≥ 10 mg/mL) and H2O (2 mg/mL with warming)
  • Utilized to induce chronic kidney injury models
  • Used in inducing Chronic Kidney Disease (CKD) models in mice and rats

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.