Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenine | 73-24-5 | 99.99% | 1 ML
SDP

Catalog No. 5000406906
Encompass_Preferred

Adenine is a purine, also known as 6-Aminopurine. It is one of the four nucleobases found in the nucleic acids DNA and RNA. Adenine plays a critical role in biochemistry, particularly in cellular respiration, as a component of ATP and cofactors like NAD and FAD, and in protein synthesis.

  • Purity: 99.99%
  • CAS number: 73-24-5
  • Molecular weight: 135.13
  • Formula: C5H5N5
  • Appearance: Solid, white to off-white
  • Soluble in DMSO (≥ 10 mg/mL)
  • Soluble in H2O (2 mg/mL; requires ultrasonic and warming to 60°C)
  • Can be used to induce chronic kidney injury models
  • Critical role in biochemistry

Catalog No. 50-004-06906 Supplier Medchemexpress LLC Supplier No. HYB015210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenine is a purine, also known as 6-Aminopurine. It is one of the four nucleobases found in the nucleic acids DNA and RNA. Adenine plays a critical role in biochemistry, particularly in cellular respiration, as a component of ATP and cofactors like NAD and FAD, and in protein synthesis.

  • Purity: 99.99%
  • CAS number: 73-24-5
  • Molecular weight: 135.13
  • Formula: C5H5N5
  • Appearance: Solid, white to off-white
  • Soluble in DMSO (≥ 10 mg/mL)
  • Soluble in H2O (2 mg/mL; requires ultrasonic and warming to 60°C)
  • Can be used to induce chronic kidney injury models
  • Critical role in biochemistry

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.