Learn More
Sigma Aldrich Fine Chemicals Biosciences Adenine 991KG
Catalog No. 502903376
Encompass
Adenine is a purine nucleobase. It is part of DNA and RNA. Adenine is also a component of cofactors (NAD fAD) and signaling molecules (cAMP).
Catalog No. 50-290-3376 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A86261KG
Adenine is a purine nucleobase. It is part of DNA and RNA. Adenine is also a component of cofactors (NAD fAD) and signaling molecules (cAMP).