Learn More
Medchemexpress LLC Adenine-d (6-aminopurine-d1) | 109923-52-6 | 99.9% | 5 MG
SDP
Catalog No. 5000407167
Encompass_Preferred
Adenine-d is a deuterium-labeled adenine, a purine nucleobase essential for DNA and RNA. It plays a crucial role in biochemistry, including cellular respiration, ATP formation, and protein synthesis. This compound can be used as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07167 Supplier Medchemexpress LLC Supplier No. HYB0152S5MG
Adenine-d is a deuterium-labeled adenine, a purine nucleobase essential for DNA and RNA. It plays a crucial role in biochemistry, including cellular respiration, ATP formation, and protein synthesis. This compound can be used as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS