Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenine-d (6-aminopurine-d1) | 109923-52-6 | MFCD06658867 | 99.9% | 136.13 | C5H4DN5 | 10 MG
SDP

Catalog No. 5000298943
Encompass_Preferred

Adenine-d (6-aminopurine-d1) is the deuterium-labeled analogue of adenine, supplied as a stable-isotope reagent for biochemical and analytical research. It is intended for metabolic labeling, mass spectrometry reference standards, and studies of nucleic acid metabolism.

  • Deuterium-labeled adenine for isotope tracing and analytical studies.
  • Chemical formula C5H4DN5.
  • Molecular weight 136.13 g/mol.
  • Purity 99.9% by specification.
  • CAS number 109923-52-6 for unambiguous identification.
  • Supplied as a powder commonly in a 10 MG package; store powder at -20°C for long-term stability.

Catalog No. 50-002-98943 Supplier Medchemexpress LLC Supplier No. HYB0152S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenine-d (6-aminopurine-d1) is the deuterium-labeled analogue of adenine, supplied as a stable-isotope reagent for biochemical and analytical research. It is intended for metabolic labeling, mass spectrometry reference standards, and studies of nucleic acid metabolism.

  • Deuterium-labeled adenine for isotope tracing and analytical studies.
  • Chemical formula C5H4DN5.
  • Molecular weight 136.13 g/mol.
  • Purity 99.9% by specification.
  • CAS number 109923-52-6 for unambiguous identification.
  • Supplied as a powder commonly in a 10 MG package; store powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.