Learn More
Selleck Chemical LLC Adenine HCl S1983-50mg
SDP
Catalog No. 503109598
Encompass_Preferred
Adenine HCl(6-Aminopurine hydrochloride) is a hydrochloride salt form of adenine which is a purine derivative and a nucleobase with a variety of roles in biochemistry Adenine HCl can be used to induce animal models of Tubulointerstitial Fibrosis
Catalog No. 50-310-9598 Supplier Selleck Chemical LLC Supplier No. S198350MG
Adenine HCl(6-Aminopurine hydrochloride) is a hydrochloride salt form of adenine which is a purine derivative and a nucleobase with a variety of roles in biochemistry Adenine HCl can be used to induce animal models of Tubulointerstitial Fibrosis