Learn More
Apexbio Technology LLC Adenine HCl 2922-28-3 1g
SDP
Catalog No. 502240300
Encompass_Preferred
Adenine HCl is a purine derivative functioning as a fundamental building block of nucleic acids It facilitates the biosynthesis of adenosine triphosphate (ATP) nicotinamide adenine dinucleotide (NAD) and flavin adenine dinucleotide (FAD) thus participating in cellular energy metabolism and redox reactions Adenine also exhibits affinity and agonistic properties at adenosine receptors modulating physiological processes In in vitro studies adenine demonstrates cytoprotective activity particularly in renal tubular cell injury models and exerts modulatory effects in vivo on lipid metabolism pathways influencing fatty acid synthesis and mitochondrial -oxidation Based on these properties adenine HCl holds research potential in cellular protection and metabolic disease models
This item is not returnable. View return policy
Catalog No. 50-224-0300 Supplier Apexbio Technology LLC Supplier No. B14711000
Adenine HCl is a purine derivative functioning as a fundamental building block of nucleic acids It facilitates the biosynthesis of adenosine triphosphate (ATP) nicotinamide adenine dinucleotide (NAD) and flavin adenine dinucleotide (FAD) thus participating in cellular energy metabolism and redox reactions Adenine also exhibits affinity and agonistic properties at adenosine receptors modulating physiological processes In in vitro studies adenine demonstrates cytoprotective activity particularly in renal tubular cell injury models and exerts modulatory effects in vivo on lipid metabolism pathways influencing fatty acid synthesis and mitochondrial -oxidation Based on these properties adenine HCl holds research potential in cellular protection and metabolic disease models