Learn More
Apexbio Technology LLC Adenine HCl 2922-28-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088182
Encompass_Preferred
Adenine HCl is a purine derivative functioning as a fundamental building block of nucleic acids It facilitates the biosynthesis of adenosine triphosphate (ATP) nicotinamide adenine dinucleotide (NAD) and flavin adenine dinucleotide (FAD) thus participating in cellular energy metabolism and redox reactions Adenine also exhibits affinity and agonistic properties at adenosine receptors modulating physiological processes In in vitro studies adenine demonstrates cytoprotective activity particularly in renal tubular cell injury models and exerts modulatory effects in vivo on lipid metabolism pathways influencing fatty acid synthesis and mitochondrial -oxidation Based on these properties adenine HCl holds research potential in cellular protection and metabolic disease models
Catalog No. 50-000-88182 Supplier Apexbio Technology LLC Supplier No. B14715.1
Adenine HCl is a purine derivative functioning as a fundamental building block of nucleic acids It facilitates the biosynthesis of adenosine triphosphate (ATP) nicotinamide adenine dinucleotide (NAD) and flavin adenine dinucleotide (FAD) thus participating in cellular energy metabolism and redox reactions Adenine also exhibits affinity and agonistic properties at adenosine receptors modulating physiological processes In in vitro studies adenine demonstrates cytoprotective activity particularly in renal tubular cell injury models and exerts modulatory effects in vivo on lipid metabolism pathways influencing fatty acid synthesis and mitochondrial -oxidation Based on these properties adenine HCl holds research potential in cellular protection and metabolic disease models