Learn More
Medchemexpress LLC Adenine (hemisulfate) | 321-30-2 | 99.9% | 1 G
SDP
Catalog No. 5000406452
Encompass_Preferred
Adenine hemisulfate (6-Aminopurine hemisulfate) is a purine, one of the four nucleobases found in the nucleic acid of DNA. It functions as a chemical component of both DNA and RNA and plays a significant role in biochemistry related to cellular respiration, forming ATP and cofactors like NAD and FAD, and protein synthesis.
  • Purine and nucleobase found in DNA and RNA
  • Chemical component of DNA and RNA
  • Plays a significant role in cellular respiration, ATP formation, NAD and FAD cofactors, and protein synthesis
  • Used to induce chronic liver injury models
  • Used for induction of chronic kidney disease (CKD) in mice and rats
Catalog No. 50-004-06452 Supplier Medchemexpress LLC Supplier No. HYB0152B1G
Adenine hemisulfate (6-Aminopurine hemisulfate) is a purine, one of the four nucleobases found in the nucleic acid of DNA. It functions as a chemical component of both DNA and RNA and plays a significant role in biochemistry related to cellular respiration, forming ATP and cofactors like NAD and FAD, and protein synthesis.
  • Purine and nucleobase found in DNA and RNA
  • Chemical component of DNA and RNA
  • Plays a significant role in cellular respiration, ATP formation, NAD and FAD cofactors, and protein synthesis
  • Used to induce chronic liver injury models
  • Used for induction of chronic kidney disease (CKD) in mice and rats