Learn More
Medchemexpress LLC Adenine (hemisulfate) | 321-30-2 | 99.9% | 1 ML
SDP
Catalog No. 5000406224
Encompass_Preferred
Adenine hemisulfate, a purine, is one of the four nucleobases in the nucleic acid of DNA. It acts as a chemical component of DNA and RNA, playing an important role in biochemistry involved in cellular respiration, the formation of both ATP and the cofactors (NAD and FAD), and protein synthesis. This product is for research use only.
  • Acts as a chemical component of DNA and RNA
  • Plays an important role in biochemistry involved in cellular respiration
  • Involved in the formation of both ATP and the cofactors (NAD and FAD)
  • Contributes to protein synthesis
  • Can be used to induce chronic liver injury models
  • Used in the induction of chronic kidney disease (CKD) models
Catalog No. 50-004-06224 Supplier Medchemexpress LLC Supplier No. HYB0152B10MM/1ML
Adenine hemisulfate, a purine, is one of the four nucleobases in the nucleic acid of DNA. It acts as a chemical component of DNA and RNA, playing an important role in biochemistry involved in cellular respiration, the formation of both ATP and the cofactors (NAD and FAD), and protein synthesis. This product is for research use only.
  • Acts as a chemical component of DNA and RNA
  • Plays an important role in biochemistry involved in cellular respiration
  • Involved in the formation of both ATP and the cofactors (NAD and FAD)
  • Contributes to protein synthesis
  • Can be used to induce chronic liver injury models
  • Used in the induction of chronic kidney disease (CKD) models