Learn More
Medchemexpress LLC Adenine hydrochloride | 2922-28-3 | 98.0% | 5 G
SDP
Catalog No. 5000407164
Encompass_Preferred
Adenine hydrochloride (6-Aminopurine hydrochloride) is a purine, and one of the four nucleobases found in the nucleic acid of DNA. It functions as a chemical component of both DNA and RNA and is involved in critical biochemical processes such as cellular respiration, the formation of ATP and cofactors (NAD and FAD), and protein synthesis.
  • A purine nucleobase found in DNA and RNA.
  • Essential for cellular respiration.
  • Contributes to the formation of ATP and cofactors (NAD, FAD).
  • Plays a role in protein synthesis.
  • For research use only.
Catalog No. 50-004-07164 Supplier Medchemexpress LLC Supplier No. HYB0152A5G
Adenine hydrochloride (6-Aminopurine hydrochloride) is a purine, and one of the four nucleobases found in the nucleic acid of DNA. It functions as a chemical component of both DNA and RNA and is involved in critical biochemical processes such as cellular respiration, the formation of ATP and cofactors (NAD and FAD), and protein synthesis.
  • A purine nucleobase found in DNA and RNA.
  • Essential for cellular respiration.
  • Contributes to the formation of ATP and cofactors (NAD, FAD).
  • Plays a role in protein synthesis.
  • For research use only.