Learn More
Medchemexpress LLC Adenine hydrochloride | 2922-28-3 | 98.0% | 500 MG
SDP
Catalog No. 5000406077
Encompass_Preferred
Adenine hydrochloride (6-Aminopurine hydrochloride), a purine, is one of the four nucleobases in the nucleic acid of DNA. It acts as a chemical component of DNA and RNA. Adenine hydrochloride also plays an important role in biochemistry involved in cellular respiration, the form of both ATP and the cofactors (NAD and FAD), and protein synthesis.
  • A purine, one of the four nucleobases in the nucleic acid of DNA.
  • Acts as a chemical component of DNA and RNA.
  • Plays an important role in biochemistry involved in cellular respiration, ATP, NAD, FAD, and protein synthesis.
  • Can be utilized to induce chronic liver injury models.
  • Used for the induction of chronic kidney disease (CKD).
Catalog No. 50-004-06077 Supplier Medchemexpress LLC Supplier No. HYB0152A500MG
Adenine hydrochloride (6-Aminopurine hydrochloride), a purine, is one of the four nucleobases in the nucleic acid of DNA. It acts as a chemical component of DNA and RNA. Adenine hydrochloride also plays an important role in biochemistry involved in cellular respiration, the form of both ATP and the cofactors (NAD and FAD), and protein synthesis.
  • A purine, one of the four nucleobases in the nucleic acid of DNA.
  • Acts as a chemical component of DNA and RNA.
  • Plays an important role in biochemistry involved in cellular respiration, ATP, NAD, FAD, and protein synthesis.
  • Can be utilized to induce chronic liver injury models.
  • Used for the induction of chronic kidney disease (CKD).