Learn More
Medchemexpress LLC Adenine (hydrochloride) | 2922-28-3 | MFCD00075782 | 98.0% | 1 ML
SDP
Catalog No. 5000406251
Encompass_Preferred
Adenine hydrochloride (6-Aminopurine hydrochloride), a purine, is one of the four nucleobases found in the nucleic acid of DNA. It functions as a chemical component of both DNA and RNA and plays a significant role in biochemistry, specifically in cellular respiration, the formation of ATP and cofactors (NAD and FAD), and protein synthesis.
  • Purine, nucleobase in DNA
  • Chemical component of DNA and RNA
  • Important in cellular respiration, ATP and cofactor formation (NAD and FAD), and protein synthesis
  • Can be used to induce chronic liver injury models
  • Used for induction of chronic kidney disease (CKD) in mice and rats
  • Appearance: solid, white to off-white color
Catalog No. 50-004-06251 Supplier Medchemexpress LLC Supplier No. HYB0152A10MM/1ML
Adenine hydrochloride (6-Aminopurine hydrochloride), a purine, is one of the four nucleobases found in the nucleic acid of DNA. It functions as a chemical component of both DNA and RNA and plays a significant role in biochemistry, specifically in cellular respiration, the formation of ATP and cofactors (NAD and FAD), and protein synthesis.
  • Purine, nucleobase in DNA
  • Chemical component of DNA and RNA
  • Important in cellular respiration, ATP and cofactor formation (NAD and FAD), and protein synthesis
  • Can be used to induce chronic liver injury models
  • Used for induction of chronic kidney disease (CKD) in mice and rats
  • Appearance: solid, white to off-white color