Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenine hydrochloride, 99.20%, MP Biomedicals™

Catalog No. ICN19460880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19460880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19460880 Supplier MP Biomedicals, Inc Supplier No. 0219460880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6055-72-7
Molecular Formula C5H8ClN5O
Molecular Weight (g/mol) 189.60
MDL Number MFCD11046306
InChI Key MYRDTAUFFBYTHA-UHFFFAOYSA-N
Synonym 7h-purin-6-amine hydrochloride hydrate, adenine hydrochloride hydrate, adenine hydrochloride hemihydrate, 1h-purin-6-amine monohydrochloride hydrate 2:1, ade hydrate hydrochloride, purine-6-ylamine, chloride, hydrate, c5h5n5.hcl.h2o, adenine hydrochloride, adeninehydrochloridehemihydrate, vitamin b4 hydrate hydrochloride
PubChem CID 24211900
IUPAC Name 7H-purin-6-amine hydrate hydrochloride
SMILES O.Cl.NC1=C2NC=NC2=NC=N1

Specifications

Color White
Assay Percent Range 0.992
Quantity 100 g
Formula Weight 171.6
Physical Form Powder
Chemical Name or Material Adenine hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.