Learn More
Medchemexpress LLC Adenine monohydrochloride hemihydrate | 6055-72-7 | MFCD11046306 | 99.9% | 189.60 | C5H8ClN5O | 1 ML
SDP
Catalog No. 5000303668
Encompass_Preferred
Adenine monohydrochloride hemihydrate is the hydrochloride salt (hemihydrate) of adenine, supplied as a solid or as a pre-made 10 mM solution in DMSO. It is used as a biochemical reagent and nucleobase source in molecular biology and biochemical assays, and is provided with supporting analytical documentation for quality control.
  • High purity (99.87%) for reliable analytical results.
  • Available as solid and 10 mM solution in DMSO for experimental flexibility.
  • White to off-white solid, easy to handle and store.
  • Supplied with COA, SDS, HPLC, and MS documentation for verification.
  • Store at 4°C and protect from light; solutions: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-003-03668 Supplier Medchemexpress LLC Supplier No. HYW01521310MM/1ML
Adenine monohydrochloride hemihydrate is the hydrochloride salt (hemihydrate) of adenine, supplied as a solid or as a pre-made 10 mM solution in DMSO. It is used as a biochemical reagent and nucleobase source in molecular biology and biochemical assays, and is provided with supporting analytical documentation for quality control.
  • High purity (99.87%) for reliable analytical results.
  • Available as solid and 10 mM solution in DMSO for experimental flexibility.
  • White to off-white solid, easy to handle and store.
  • Supplied with COA, SDS, HPLC, and MS documentation for verification.
  • Store at 4°C and protect from light; solutions: -80°C (6 months) or -20°C (1 month).