Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenine phosphate | 70700-30-0 | 100.0% | 233.12 | 5 G
SDP

Catalog No. 5000452057
Encompass_Preferred

Adenine phosphate (6-Aminopurine phosphate) can stimulate leukocyte proliferation. It is a biological material or organic compound widely used in life science research.

  • Appears as a solid
  • Color is white to off-white
  • Can be stored at room temperature, dry and cool
  • Soluble in DMSO (11.67 mg/mL)

Catalog No. 50-004-52057 Supplier Medchemexpress LLC Supplier No. HYW0096325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenine phosphate (6-Aminopurine phosphate) can stimulate leukocyte proliferation. It is a biological material or organic compound widely used in life science research.

  • Appears as a solid
  • Color is white to off-white
  • Can be stored at room temperature, dry and cool
  • Soluble in DMSO (11.67 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.