Learn More
Medchemexpress LLC Adenine (Standard) | 73-24-5 | 99.9% | 100 MG
SDP
Catalog No. 5000406265
Encompass_Preferred
Adenine (Standard) is the analytical standard of Adenine, intended for research and analytical applications. Adenine (6-Aminopurine), a purine, is one of the four nucleobases in the nucleic acid of DNA. It acts as a chemical component of DNA and RNA and plays an important role in biochemistry involved in cellular respiration, the form of both ATP and the cofactors (NAD and FAD), and protein synthesis. The compound is an analytical standard, supplied as a reference standard assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-06265 Supplier Medchemexpress LLC Supplier No. HYB0152R100MG
Adenine (Standard) is the analytical standard of Adenine, intended for research and analytical applications. Adenine (6-Aminopurine), a purine, is one of the four nucleobases in the nucleic acid of DNA. It acts as a chemical component of DNA and RNA and plays an important role in biochemistry involved in cellular respiration, the form of both ATP and the cofactors (NAD and FAD), and protein synthesis. The compound is an analytical standard, supplied as a reference standard assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications