Learn More
Medchemexpress LLC Adenine (Standard) | 73-24-5 | 99.91% | 25 MG
SDP
Catalog No. 5000406525
Encompass_Preferred
Adenine (Standard) is the analytical standard of Adenine, intended for research and analytical applications. Adenine (6-Aminopurine), a purine, is one of the four nucleobases found in the nucleic acid of DNA. It acts as a chemical component of DNA and RNA and plays an important role in biochemistry involved in cellular respiration, forming both ATP and cofactors (NAD and FAD), and protein synthesis.
  • Intended for research and analytical applications
  • A purine and one of the four nucleobases found in DNA and RNA
  • Acts as a chemical component of DNA and RNA
  • Plays an important role in biochemistry involved in cellular respiration, forming both ATP and cofactors (NAD and FAD), and protein synthesis
  • Used as a reference standard for assay
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS
Catalog No. 50-004-06525 Supplier Medchemexpress LLC Supplier No. 25MGHYB0152R
Adenine (Standard) is the analytical standard of Adenine, intended for research and analytical applications. Adenine (6-Aminopurine), a purine, is one of the four nucleobases found in the nucleic acid of DNA. It acts as a chemical component of DNA and RNA and plays an important role in biochemistry involved in cellular respiration, forming both ATP and cofactors (NAD and FAD), and protein synthesis.
  • Intended for research and analytical applications
  • A purine and one of the four nucleobases found in DNA and RNA
  • Acts as a chemical component of DNA and RNA
  • Plays an important role in biochemistry involved in cellular respiration, forming both ATP and cofactors (NAD and FAD), and protein synthesis
  • Used as a reference standard for assay
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS