Learn More
Bioworld Adenine Sulfate, 100 g
Supplier:  Bioworld 401000402
Encompass_Preferred
Technical Information:ACS GradeSynonym: Adenine hemisulfate; 6-Aminopurine hemisulfate salt; 1H-Purin-6-amine sulfateAppearance: White to light yellow crystal powderBoiling Point : 553.5 ¡ãC at 760 mmHgHS Code : 293359900Melting Point: 285 ¡ãC (dec.)(lit.)RTECS Number: AU7140000
Catalog No. 50-255-198