Learn More
Selleck Chemical LLC Adenine sulfate S1982-50mg
SDP
Catalog No. 503109797
Encompass_Preferred
Adenine sulfate(Adenine hemisulfate) is a sulfate salt form of adenine which is a purine derivative and a nucleobase with a variety of roles in biochemistry Adenine sulfate can be used to induce animal models of Tubulointerstitial Fibrosis
Catalog No. 50-310-9797 Supplier Selleck Chemical LLC Supplier No. S198250MG
Adenine sulfate(Adenine hemisulfate) is a sulfate salt form of adenine which is a purine derivative and a nucleobase with a variety of roles in biochemistry Adenine sulfate can be used to induce animal models of Tubulointerstitial Fibrosis