Learn More
Apexbio Technology LLC Adenine sulfate 321-30-2 10mM (in 1mL H2O)
SDP
Catalog No. 5000087646
Encompass_Preferred
Adenine sulfate (CAS 321-30-2) is a purine nucleobase derivative used as a biochemical substrate in research investigating purine metabolism and nucleotide biosynthesis disorders Adenine sulfate is frequently employed in experimental models of renal pathophysiology particularly for inducing chronic kidney disease via renal tubular injury In in vitro studies inhibition effects involving adenine compounds have been reported typically with IC50 values in micromolar to millimolar ranges depending on enzymatic target specificity and assay conditions Based on these properties adenine sulfate holds research potential in studies of cellular energetics metabolic regulation and renal disease mechanisms
Catalog No. 50-000-87646 Supplier Apexbio Technology LLC Supplier No. B14695.1
Adenine sulfate (CAS 321-30-2) is a purine nucleobase derivative used as a biochemical substrate in research investigating purine metabolism and nucleotide biosynthesis disorders Adenine sulfate is frequently employed in experimental models of renal pathophysiology particularly for inducing chronic kidney disease via renal tubular injury In in vitro studies inhibition effects involving adenine compounds have been reported typically with IC50 values in micromolar to millimolar ranges depending on enzymatic target specificity and assay conditions Based on these properties adenine sulfate holds research potential in studies of cellular energetics metabolic regulation and renal disease mechanisms