Learn More
Alkali Scientific Adenine Sulfate [Adenine Hemisulfate Salt], 25 Grams
SDP
Catalog No. 5000661531
Encompass_Preferred
Adenine Sulfate, also known as Adenine Hemisulfate Salt, is a high-purity chemical reagent available in a 25-gram package. This compound is a purine base and an essential component of nucleotides, playing a crucial role in DNA and RNA synthesis. Adenine is widely used in molecular biology research, particularly in studies involving genetic material, polymerase chain reaction (PCR), and gene expression analysis. Its high purity ensures consistent and reliable results in experimental applications. Adenine sulfate is also used in studies on cellular metabolism, enzymatic activity, and in vitro synthesis of nucleic acids for genetic research and diagnostics.
Catalog No. 50-006-61531 Supplier Alkali Scientific Supplier No. ADE001CP
Adenine Sulfate, also known as Adenine Hemisulfate Salt, is a high-purity chemical reagent available in a 25-gram package. This compound is a purine base and an essential component of nucleotides, playing a crucial role in DNA and RNA synthesis. Adenine is widely used in molecular biology research, particularly in studies involving genetic material, polymerase chain reaction (PCR), and gene expression analysis. Its high purity ensures consistent and reliable results in experimental applications. Adenine sulfate is also used in studies on cellular metabolism, enzymatic activity, and in vitro synthesis of nucleic acids for genetic research and diagnostics.