Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenine, Synthetic, Spectrum™ Chemical
SDP

Catalog No. 18620159 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-159 1 kg
18-620-156 5 g
18-620-157 25 g
18-620-158 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-159 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AD1051KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73-24-5

Specifications

Packaging Drum
Quantity 1 kg
Molecular Weight (g/mol) 135.14
Percent Purity 98 to 102%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Adenine, Synthetic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.