Learn More
Cayman Chemical Adenosine 3',5'-diphosphate (sodium salt), Purity: ≥95%, CAS: 75431-54-8, 10 mg
SDP
Catalog No. 503015887
Encompass_Preferred
An adenine nucleotide containing two phosphate groups; a product of PAPS; has been used to study the kinetic properties and structure of SULTs
Catalog No. 50-301-5887 Supplier Cayman Chemical Supplier No. 2426510 MG
An adenine nucleotide containing two phosphate groups; a product of PAPS; has been used to study the kinetic properties and structure of SULTs