Learn More
Cayman Chemical Adenosine 3'-monophosphate (sodium salt hydrate), Purity: ≥98%, 50 mg
SDP
Catalog No. 502934066
Encompass_Preferred
A nucleotide and metabolite; formed via hydrolysis of 2',3'-cAMP by metal-dependent phosphodiesterases; reduces proliferation of rat PGVSMCs and GMCs; metabolic intermediate in the biosynthesis of adenosine
Catalog No. 50-293-4066 Supplier Cayman Chemical Supplier No. 2625650 MG
A nucleotide and metabolite; formed via hydrolysis of 2',3'-cAMP by metal-dependent phosphodiesterases; reduces proliferation of rat PGVSMCs and GMCs; metabolic intermediate in the biosynthesis of adenosine