Learn More
Cayman Chemical Adenosine 5'-diphosphate, Purity: ≥95%, CAS: 58-64-0, 250 mg
SDP
Catalog No. 502965865
Encompass_Preferred
A central component of energy storage, metabolism, and signal transduction in vivo; a precursor for ATP and agonist of purinergic receptors
Catalog No. 50-296-5865 Supplier Cayman Chemical Supplier No. 16778250 MG
A central component of energy storage, metabolism, and signal transduction in vivo; a precursor for ATP and agonist of purinergic receptors