Learn More
Cayman Chemical AdenosIn 5-phosphosulfate 10mg
SDP
Catalog No. 502985135
Encompass_Preferred
An inhibitor of ATP sulfurylase (Ki = 18, 2500, and 1500 nM in humans, S. cerevisiae, P. chrysogenum, respectively) and APS kinase (Ki = 47.5 μM); prevents sulfation
Catalog No. 50-298-5135 Supplier Cayman Chemical Supplier No. 2122610 MG
An inhibitor of ATP sulfurylase (Ki = 18, 2500, and 1500 nM in humans, S. cerevisiae, P. chrysogenum, respectively) and APS kinase (Ki = 47.5 μM); prevents sulfation