Learn More
Cayman Chemical Adenosine 5'-(γ-thio)-triphosphate (lithium salt), Purity: ≥90%, CAS: 93839-89-5, 1 mg
SDP
Catalog No. 502970702
Encompass_Preferred
A stable analog of ATP that acts as a potent agonist of G protein-coupled P2Y2 and P2Y11 receptors (pEC50 = 5.52 for P2Y11)
Catalog No. 50-297-0702 Supplier Cayman Chemical Supplier No. 149571 MG
A stable analog of ATP that acts as a potent agonist of G protein-coupled P2Y2 and P2Y11 receptors (pEC50 = 5.52 for P2Y11)