Learn More
Cayman Chemical AdenosIn 5- -thIo-trIphos 5mg
SDP
Catalog No. 502971132
Encompass_Preferred
A stable analog of ATP that acts as a potent agonist of G protein-coupled P2Y2 and P2Y11 receptors (pEC50 = 5.52 for P2Y11)
Catalog No. 50-297-1132 Supplier Cayman Chemical Supplier No. 149575 MG
A stable analog of ATP that acts as a potent agonist of G protein-coupled P2Y2 and P2Y11 receptors (pEC50 = 5.52 for P2Y11)